Anti-TAOK2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TAOK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-944
- 77844-943
- Antigen Symbol:
- TAOK2
- TAOK2
- Antigen Synonyms:
- 1110033K02Rik|B230344N16|hKFC C|hKFC-C|KIAA0881|Kinase from chicken homolog C|MAP3K17|mKIAA0881|Prostate derived STE20 like kinase 1|Prostate derived STE20 like kinase PSK|Prostate derived sterile 20 like kinase 1|Prostate-derived STE20-like kinase 1|PSK 1|PSK|PSK-1|PSK1|PSK1 beta|Serine/threonine protein kinase TAO2|Serine/threonine-protein kinase TAO2|TAO 1|TAO 2|TAO kinase 2|TAO1|TAO2|TAOK 2|TAOK2_HUMAN|Thousand and one amino acid protein 2|Thousand and one amino acid protein kinase|UNQ2971/PRO7431
- 1110033K02Rik|B230344N16|hKFC C|hKFC-C|KIAA0881|Kinase from chicken homolog C|MAP3K17|mKIAA0881|Prostate derived STE20 like kinase 1|Prostate derived STE20 like kinase PSK|Prostate derived sterile 20 like kinase 1|Prostate-derived STE20-like kinase 1|PSK 1|PSK|PSK-1|PSK1|PSK1 beta|Serine/threonine protein kinase TAO2|Serine/threonine-protein kinase TAO2|TAO 1|TAO 2|TAO kinase 2|TAO1|TAO2|TAOK 2|TAOK2_HUMAN|Thousand and one amino acid protein 2|Thousand and one amino acid protein kinase|UNQ2971/PRO7431
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UL54
- Q9UL54
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/threonine-protein kinase TAO2 protein (719-880AA)
- Recombinant Human Serine/threonine-protein kinase TAO2 protein (719-880AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice