Anti-FARP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: FARP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-381
- 77846-380
- Antigen Symbol:
- FARP1
- FARP1
- Antigen Synonyms:
- CDEP|Chondrocyte derived ezrin like protein|Chondrocyte-derived ezrin-like protein|FARP1_HUMAN|FERM RhoGEF and pleckstrin domain containing protein 1|FERM|FERM RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived)|ARHGEF|and pleckstrin domain protein 1|PH domain-containing family C member 2|Pleckstrin homology domain-containing family C member 2|PLEKHC2|PPP1R75|protein phosphatase 1|regulatory subunit 75|RhoGEF and pleckstrin domain-containing protein 1
- CDEP|Chondrocyte derived ezrin like protein|Chondrocyte-derived ezrin-like protein|FARP1_HUMAN|FERM RhoGEF and pleckstrin domain containing protein 1|FERM|FERM RhoGEF (ARHGEF) and pleckstrin domain protein 1 (chondrocyte-derived)|ARHGEF|and pleckstrin domain protein 1|PH domain-containing family C member 2|Pleckstrin homology domain-containing family C member 2|PLEKHC2|PPP1R75|protein phosphatase 1|regulatory subunit 75|RhoGEF and pleckstrin domain-containing protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y4F1
- Q9Y4F1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human FERM, ARHGEF and pleckstrin domain-containing protein 1 protein (1-300AA)
- Recombinant Human FERM, ARHGEF and pleckstrin domain-containing protein 1 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice