Anti-DEFB126 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77854-106
New Product
- Antigen Symbol:DEFB126
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pongo pygmaeus
- Size:50 µg
- Form:Liquid
- Gene ID:A4H244
- Antigen Synonyms:DEFB126Beta-defensin 126|Defensin|beta 126
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Pongo pygmaeus Beta-defensin 126 protein (21-63AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: DEFB126
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pongo pygmaeus