Anti-C5 Rabbit Recombinant Polyclonal Antibody
77852-125
New Product
- Antigen Symbol:C5
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P01031
- Antigen Synonyms:Anaphylatoxin C5a analog|C3 and PZP-like alpha-2-macroglobulin domain-containing protein 4|C5|C5a anaphylatoxin|C5a|C5b|CO5_HUMAN|Complement C5 alpha'' chain|Complement C5|Complement component C5|CPAMD4|prepro-C5
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Complement C5 protein (678-751AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: C5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human