Anti-CACNA1H Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-251
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HUS1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O60921
- Antigen Synonyms:Checkpoint protein HUS1|hHUS1|HUS1 (S. pombe) checkpoint homolog|Hus1|HUS1 checkpoint homolog (S. pombe)|HUS1 Checkpoint Protein|HUS1 protein|HUS1+ - like protein|HUS1_HUMAN|Hydroxyurea-sensitive 1|S. pombe|homolog of
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Checkpoint protein HUS1 protein (2-280AA)
- Purification:>95%, Protein G purified
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human