Anti-MAD2L1BP Rabbit Polyclonal Antibody
Catalog # 77846-795
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAD2L1BP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:Q15013
- Antigen Synonyms:Caught by MAD2 protein|CMT2|dJ261G23.1|KIAA0110|MAD2L1 binding protein|MAD2L1 binding protein isoform 2|MAD2L1-binding protein|Mad2l1bp|MD2BP_HUMAN|MGC11282|OTTHUMP00000016496|RP1 261G23.6
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from N-terminal of Human MAD2L1BP.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:3000
Type:
Antigen: MAD2L1BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human