Anti-TRAP1 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA:1:2000 to 1:10000, WB: 1:1000 to 1:5000, IHC: 1:100 to 1:200
Type:
Antigen: TRAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- 77848-251
- 77848-252
- Antigen Symbol:
- TRAP1
- TRAP1
- Antigen Synonyms:
- Heat shock protein 75 kDa|Heat shock protein 75 kDa, mitochondrial|HSP 75|HSP75|HSP90L|mitochondrial|TNF receptor associated protein 1|TNFR-associated protein 1|TRAP-1|TRAP1_HUMAN|Tumor necrosis factor type 1 receptor-associated protein
- Heat shock protein 75 kDa|Heat shock protein 75 kDa, mitochondrial|HSP 75|HSP75|HSP90L|mitochondrial|TNF receptor associated protein 1|TNFR-associated protein 1|TRAP-1|TRAP1_HUMAN|Tumor necrosis factor type 1 receptor-associated protein
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q12931
- Q12931
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human TRAP1
- Fusion protein of Human TRAP1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice