Anti-SLC10A7 Rabbit Polyclonal Antibody
77848-354
New Product
- Antigen Symbol:SLC10A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q0GE19
- Antigen Synonyms:C4orf13|P7|PSEC0051|Sodium/bile acid cotransporter 7|Na(+)/bile acid cotransporter 7|Solute carrier family 10 member 7
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human SLC10A7.
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: SLC10A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human