Anti-ADAM10 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77850-725
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ADAM10
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O14672
- Antigen Synonyms:A disintegrin and metalloprotease domain 10|A disintegrin and metalloproteinase domain 10|AD 10|AD10|AD18|ADA10_HUMAN|ADAM 10|ADAM metallopeptidase domain 10|CD 156c|CD156c|CD156c antigen|CDw156|disintegrin and metalloproteinase domain containing protein 10|Disintegrin and metalloproteinase domain-containing protein 10|HsT 18717|HsT18717|Kuz|Kuzbanian|Kuzbanian protein homolog|Drosophila|homolog of|MADM|Mammalian disintegrin metalloprotease|Mammalian disintegrin-metalloprotease|RAK
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Disintegrin and metalloproteinase domain-containing protein 10 protein (214-504AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ADAM10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human