Anti-DYNC1LI1 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC:1:200 to 1:500
Type:
Antigen: DYNC1LI1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-224
- 77846-223
- Antigen Symbol:
- DYNC1LI1
- DYNC1LI1
- Antigen Synonyms:
- Cytoplasmic dynein 1 light intermediate chain 1|cytosolic|DC1L1_HUMAN|DLC-A|DNCLI1|Dynein cytoplasmic 1 light intermediate chain 1|Dynein cytoplasmic light intermediate polypeptide 1|Dynein light chain A|Dynein light intermediate chain 1|Dynein light intermediate chain 1 cytosolic|FLJ10219|LIC1
- Cytoplasmic dynein 1 light intermediate chain 1|cytosolic|DC1L1_HUMAN|DLC-A|DNCLI1|Dynein cytoplasmic 1 light intermediate chain 1|Dynein cytoplasmic light intermediate polypeptide 1|Dynein light chain A|Dynein light intermediate chain 1|Dynein light intermediate chain 1 cytosolic|FLJ10219|LIC1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6G9
- Q9Y6G9
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Cytoplasmic dynein 1 light intermediate chain 1 protein (167-233AA)
- Recombinant Human Cytoplasmic dynein 1 light intermediate chain 1 protein (167-233AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice