Anti-DUSP8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000, IHC: 1:50 to 1:200
Type:
Antigen: DUSP8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-624
- 77848-623
- Antigen Symbol:
- DUSP8
- DUSP8
- Antigen Synonyms:
- C11orf81|Chromosome 11 open reading frame 81|Deubiquitinating enzyme 8|Dual Specificity Phosphatase 8|Dual specificity phosphatase M3/6|Dual specificity protein phosphatase 8|Dual specificity protein phosphatase hVH 5|Dual specificity protein phosphatase hVH-5|DUS8|DUS8_HUMAN|EC 3.1.3.16|EC 3.1.3.48|FLJ42476|FLJ42958|H1 phosphatase vaccinia virus homolog|H1 phosphatase|vaccinia virus|homolog of|HB5|hUBPy|HVH5|HVH8|M3/6|mouse|Neuronal tyrosine threonine phosphatase 1|Serine/threonine specific protein phosphatase|Ubiquitin carboxyl terminal hydrolase 8|Ubiquitin specific processing protease 8|Ubiquitin thiolesterase 8|VH5
- C11orf81|Chromosome 11 open reading frame 81|Deubiquitinating enzyme 8|Dual Specificity Phosphatase 8|Dual specificity phosphatase M3/6|Dual specificity protein phosphatase 8|Dual specificity protein phosphatase hVH 5|Dual specificity protein phosphatase hVH-5|DUS8|DUS8_HUMAN|EC 3.1.3.16|EC 3.1.3.48|FLJ42476|FLJ42958|H1 phosphatase vaccinia virus homolog|H1 phosphatase|vaccinia virus|homolog of|HB5|hUBPy|HVH5|HVH8|M3/6|mouse|Neuronal tyrosine threonine phosphatase 1|Serine/threonine specific protein phosphatase|Ubiquitin carboxyl terminal hydrolase 8|Ubiquitin specific processing protease 8|Ubiquitin thiolesterase 8|VH5
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13202
- Q13202
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human DUSP8
- Synthetic peptide of Human DUSP8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice