Anti-H2AFJ Rabbit Recombinant Polyclonal Antibody
Catalog # 77856-675
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:H2AFJ
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9BTM1
- Antigen Synonyms:H2AFJ| Histone H2A.J| H2a/j
- Storage Buffer:0.1 mol/L NaCl, 15 mmol/L NaN₃, pH 7.2.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Histone H2A.J protein (2-129AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: H2AFJ
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse