Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Human Claudin 6 (from HEK293 cells)
Recombinant Human Claudin 6 (from HEK293 cells)
Recombinant Human Claudin 6 (from HEK293 cells)
Supplier:
ANTIBODIES.COM LLC
Certificates
Expand image
Recombinant Human Claudin 6 (from HEK293 cells)
Supplier:
ANTIBODIES.COM LLC
About this item