Anti-MC4R Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: MC4R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-977
- 77847-978
- Antigen Symbol:
- MC4R
- MC4R
- Antigen Synonyms:
- MC4 Receptor|MC4-R|MC4R_HUMAN|Melanocortin 4 receptor|Melanocortin receptor 4|MGC126851|MGC138197
- MC4 Receptor|MC4-R|MC4R_HUMAN|Melanocortin 4 receptor|Melanocortin receptor 4|MGC126851|MGC138197
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P32245
- P32245
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human MC4R
- Synthetic peptide of Human MC4R
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice