Anti-SLC12A4 Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: SLC12A4
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-049
- 77845-050
- Antigen Symbol:
- SLC12A4
- SLC12A4
- Antigen Synonyms:
- Electroneutral potassium chloride cotransporter 1|Electroneutral potassium-chloride cotransporter 1|Erythroid K Cl cotransporter 1|Erythroid K-Cl cotransporter 1|FLJ40489|hKCC 1|hKCC1|KCC 1|KCC1|Potassium transport protein|Potassium/chloride cotransporter 1|S12A4_HUMAN|Solute carrier family 12 (potassium/chloride transporters) member 4|Solute carrier family 12 member 4
- Electroneutral potassium chloride cotransporter 1|Electroneutral potassium-chloride cotransporter 1|Erythroid K Cl cotransporter 1|Erythroid K-Cl cotransporter 1|FLJ40489|hKCC 1|hKCC1|KCC 1|KCC1|Potassium transport protein|Potassium/chloride cotransporter 1|S12A4_HUMAN|Solute carrier family 12 (potassium/chloride transporters) member 4|Solute carrier family 12 member 4
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UP95
- Q9UP95
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Solute carrier family 12 member 4 protein (601-900AA)
- Recombinant Human Solute carrier family 12 member 4 protein (601-900AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice