Anti-DCAF6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: DCAF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77849-511
- 77849-510
- Antigen Symbol:
- DCAF6
- DCAF6
- Antigen Synonyms:
- 1200006M05Rik|Androgen receptor complex associated protein|Androgen receptor complex-associated protein|ARCAP|DCAF6_HUMAN|DDB1 and CUL4 associated factor 6|DDB1- and CUL4-associated factor 6|IQ motif and WD repeat-containing protein 1|IQ motif and WD repeats 1|IQWD1|MSTP055|NRIP|Nuclear receptor interaction protein|RP4 745I14.1
- 1200006M05Rik|Androgen receptor complex associated protein|Androgen receptor complex-associated protein|ARCAP|DCAF6_HUMAN|DDB1 and CUL4 associated factor 6|DDB1- and CUL4-associated factor 6|IQ motif and WD repeat-containing protein 1|IQ motif and WD repeats 1|IQWD1|MSTP055|NRIP|Nuclear receptor interaction protein|RP4 745I14.1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q58WW2
- Q58WW2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human DCAF6
- Synthetic peptide of Human DCAF6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice