Anti-MAP2K4 Rabbit Recombinant Polyclonal Antibody
Catalog # 77859-368
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MAP2K4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P45985
- Antigen Synonyms:c Jun N terminal kinase kinase 1|C-JUN N-terminal kinase kinase 1|Dual specificity mitogen activated protein kinase kinase 4|Dual specificity mitogen-activated protein kinase kinase 4|JNK Activated Kinase 1|JNK activating kinase 1|JNK-activating kinase 1|JNKK|JNKK1|MAP kinase kinase 4|MAPK ERK kinase 4|MAPK/ERK kinase 4|MAPKK 4|MAPKK4|MEK 4|MEK4|Mitogen activated protein kinase kinase 4|MKK 4|MKK4|MP2K4_HUMAN|PRKMK4|SAPK ERK kinase 1|SAPK/ERK kinase 1|SAPKK 1|SAPKK1|SEK1|SERK1|SKK1|Stress activated protein kinase kinase 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dual specificity mitogen-activated protein kinase kinase 4 protein (259-398AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:100 to 1:500, IF: 1:50 to 1:500
Type:
Antigen: MAP2K4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human