Anti-lacZ Rabbit Recombinant Polyclonal Antibody
Catalog # 77856-061
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:NR1H2
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P55055
- Antigen Synonyms:Liver X nuclear receptor beta|Liver X receptor beta|LX receptor beta|LXR b|LXR beta|LXRB|NER|NER I|NER1|NR1H 2|NR1H2|NR1H2_HUMAN|Nuclear orphan receptor LXR beta|Nuclear receptor NER|Nuclear receptor subfamily 1 group H member 2|OR-1|Oxysterols receptor LXR-beta|RIP15|Steroid hormone nuclear receptor NER|Ubiquitously-expressed nuclear receptor|UNR
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Oxysterols receptor LXR-beta protein (1-461AA)
- Purification:>95%, Protein G purified
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human