Anti-TRPV2 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77846-597
- 77846-596
- Antigen Symbol:
- TRPV2
- TRPV2
- Antigen Synonyms:
- MGC12549|Osm 9 like TRP channel 2|Osm-9-like TRP channel 2|OTRPC2|Sac2b|Stretch activated channel 2B|Transient receptor potential cation channel subfamily V member 2|TRPV2|TRPV2_HUMAN|Vanilloid receptor like protein 1|Vanilloid receptor-like protein 1|VRL 1|VRL|VRL-1|VRL1
- MGC12549|Osm 9 like TRP channel 2|Osm-9-like TRP channel 2|OTRPC2|Sac2b|Stretch activated channel 2B|Transient receptor potential cation channel subfamily V member 2|TRPV2|TRPV2_HUMAN|Vanilloid receptor like protein 1|Vanilloid receptor-like protein 1|VRL 1|VRL|VRL-1|VRL1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y5S1
- Q9Y5S1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Transient receptor potential cation channel subfamily V member 2 protein (1-117AA)
- Recombinant Human Transient receptor potential cation channel subfamily V member 2 protein (1-117AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice