Anti-CHML Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77853-079
New Product
- Antigen Symbol:CHML
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P26374
- Antigen Synonyms:CHM like|Chml|Choroideraemia like protein|Choroideraemia-like protein|Choroideremia like (Rab escort protein 2)|Choroideremia like|Choroideremia like protein|Rab escort protein 2|Rab proteins geranylgeranyltransferase component A 2|RAE2_HUMAN|REP 2|REP-2|REP2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Rab proteins geranylgeranyltransferase component A 2 protein (480-656AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CHML
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human