Anti-KIF22 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:10000, IHC: 1:50 to 1:200
Type:
Antigen: KIF22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-077
- 77849-076
- Antigen Symbol:
- KIF22
- KIF22
- Antigen Synonyms:
- A 328A3.2|KID|KIF 22|KIF22_HUMAN|Kinesin family member 22|Kinesin like 4|Kinesin like DNA binding protein|Kinesin like DNA binding protein pseudogene|Kinesin like protein 4|Kinesin like protein Kinesin-like DNA-binding protein|Kinesin-like protein 4|Kinesin-like protein KNSL 4|KNSL4|OBP 1|OBP 2|OBP|OBP1|OBP2|Origin of plasmid DNA replication binding protein|OriP binding protein|OTTHUMP00000123406|SEMDJL2
- A 328A3.2|KID|KIF 22|KIF22_HUMAN|Kinesin family member 22|Kinesin like 4|Kinesin like DNA binding protein|Kinesin like DNA binding protein pseudogene|Kinesin like protein 4|Kinesin like protein Kinesin-like DNA-binding protein|Kinesin-like protein 4|Kinesin-like protein KNSL 4|KNSL4|OBP 1|OBP 2|OBP|OBP1|OBP2|Origin of plasmid DNA replication binding protein|OriP binding protein|OTTHUMP00000123406|SEMDJL2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q14807
- Q14807
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human KIF22
- Synthetic peptide of Human KIF22
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice