Anti-SNX12 Rabbit Recombinant polyclonal Antibody
Catalog # 77844-513
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SNX12
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9UMY4
- Antigen Synonyms:2610001F05Rik|AW045757|MGC118982|MGC118983|RGD1565585|RP23-356J7.1|SDP8|SNX 12|SNX12_HUMAN|Sorting nexin 12|Sorting nexin-12
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human Sorting nexin-12 protein (2-172AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500-1:5000, IHC:1:500-1:1000, IF:1:200-1:500, IP:1:200-1:2000
Type:
Antigen: SNX12
Clonality: Recombinant polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human