Anti-ENTPD7 Rabbit Recombinant Polyclonal Antibody
Catalog # 77854-765
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ENTPD7
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NQZ7
- Antigen Synonyms:DKFZp667O124|Ectonucleoside triphosphate diphosphohydrolase 7|ENTP7_HUMAN|ENTPD 7|Entpd7|LALP 1|LALP1|Lysosomal apyrase like protein 1|Lysosomal apyrase-like protein 1|NTPDase 7|NTPDase7
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Ectonucleoside triphosphate diphosphohydrolase 7 protein (51-350AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:2000 to 1:10000, IF: 1:50 to 1:200
Type:
Antigen: ENTPD7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse