Anti-CEBPB Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-965
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CEBPB
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P17676
- Antigen Synonyms:AGP/EBP|C EBP beta|C/EBP beta|C/EBP related protein 2|CCAAT Enhancer Binding Protein beta|CCAAT/enhancer-binding protein beta|CEBPB|CEBPB_HUMAN|CRP2|IL 6DBP|IL6DBP|Interleukin 6 dependent binding protein|LAP|Liver activator protein|Liver enriched transcriptional activator|NF IL6|NFIL6|Nuclear factor NF IL6|Nuclear factor NF-IL6|SF B|SFB|Silencer factor B|TCF-5|TCF5|Transcription factor 5
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human CCAAT/enhancer-binding protein beta protein (41-270AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CEBPB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human