Anti-SSR3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-018
- 77845-019
- Antigen Symbol:
- SSR3
- SSR3
- Antigen Synonyms:
- Signal sequence receptor gamma|Signal sequence receptor subunit gamma|SSR-gamma|Ssr3|SSRG_HUMAN|Translocon associated protein gamma subunit|Translocon-associated protein subunit gamma|TRAP complex gamma subuni|TRAP-gamma|TRAPG
- Signal sequence receptor gamma|Signal sequence receptor subunit gamma|SSR-gamma|Ssr3|SSRG_HUMAN|Translocon associated protein gamma subunit|Translocon-associated protein subunit gamma|TRAP complex gamma subuni|TRAP-gamma|TRAPG
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UNL2
- Q9UNL2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Translocon-associated protein subunit gamma protein (77-135AA)
- Recombinant Human Translocon-associated protein subunit gamma protein (77-135AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together