Anti-B3GALT4 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-737
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:B3GALT4
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:O96024
- Antigen Synonyms:3-galactosyltransferase 4|3-galactosyltransferase|3-GalTase 4|b3Gal-T4|B3GALT4|B3GT4_HUMAN|Beta 1,3 galactosyltransferase 4|Beta 1,3 GalTase 4|Beta 3 galactosyltransferase 4|Beta-1|Beta3Gal-T4|Beta3GalT4|Gal T2|GAL-T2|GALT2|GalT4|Ganglioside galactosyltransferase|OTTHUMP00000029128|UDP Gal:betaGlcNAc beta 1,3 galactosyltransferase|polypeptide 4|UDP galactose:beta N acetyl galactosamine beta 1,3 galactosyltransferase|UDP-galactose:beta-N-acetyl-galactosamine-beta-1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-1,3-galactosyltransferase 4 protein (42-266AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200
Type:
Antigen: B3GALT4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse