Anti-CPB2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-458
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CPB2
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q96IY4
- Antigen Synonyms:Carboxypeptidase B2|EC 3.4.17.20|Carboxypeptidase U|CPU|Plasma carboxypeptidase B|pCPB|Thrombin-activable fibrinolysis inhibitor|TAFI
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Carboxypeptidase B2 protein (225-372AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CPB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human