Anti-PEAK1 Rabbit Polyclonal Antibody
Catalog # 77849-533
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PEAK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:liquid
- Gene ID:Q9H792
- Antigen Synonyms:FLJ21140|FLJ34483|KIAA2002|NKF3 kinase family member|PEAK1|PEAK1_HUMAN|pseudopodium enriched atypical kinase|Pseudopodium-enriched atypical kinase 1|SGK269|Sugen kinase 269|Tyrosine-protein kinase SgK269
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from N-terminal of Human SGK269
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: PEAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human