Anti-ANXA3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-177
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ANXA3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P12429
- Antigen Synonyms:2-cyclic phosphate 2-phosphohydrolase|35-alpha calcimedin|Annexin A3|Annexin III|Annexin-3|ANX3|ANXA 3|ANXA3_HUMAN|Calcimedin 35 alpha|Inositol 1 2 cyclic phosphate 2 phosphohydrolase|Inositol 1|Lipocortin III|OTTHUMP00000160701|OTTHUMP00000219237|OTTHUMP00000219238|PAP III|PAP-III|Placental anticoagulant protein III
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Annexin A3 protein (5-322AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:10 to 1:100
Type:
Antigen: ANXA3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human