Anti-CES1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-995
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CES1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P23141
- Antigen Synonyms:ACAT|Acyl coenzyme A cholesterol acyltransferase|Acyl-coenzyme A:cholesterol acyltransferase|Brain carboxylesterase hBr1|Carboxyesterase ES-3|Carboxylesterase|Carboxylesterase 1 (monocyte/macrophage serine esterase 1)|Carboxylesterase 1|Carboxylesterase 1 deficiency|included|Carboxylesterase 2|formerly|CE 1|CEH|Ces-1|CES1|CES2|CESDD1|Cholesterol ester hydrolase|neutral|macrophage-derived|Cholesteryl ester hydrolase|Cocaine carboxylesterase|EC 3.1.1.1|Egasyn|ES-HTEL|ES-x|Es22|EST1_HUMAN|Esterase 22|Esterase|hCE 1|HMSE|HMSE1|Liver carboxylesterase 1|Liver carboxylesterase 3|Methylumbelliferyl acetate deacetylase 1|MGC117365|MGC156521|Monocyte carboxylesterase deficiency|Monocyte esterase deficiency|Monocyte/macrophage serine esterase|PCE-1|pI 5.5 esterase|Proline-beta-naphthylamidase|REH|Retinyl ester hydrolase|Serine esterase 1|Ses-1|SES1|TGH|Triacylglycerol hydrolase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Liver carboxylesterase 1 protein (291-566AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: CES1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse