Anti-ALDH7A1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-057
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ALDH7A1
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P49419
- Antigen Synonyms:26g turgor protein homolog|AL7A1_HUMAN|Aldehyde dehydrogenase 7 A1|Aldehyde dehydrogenase 7 family|member A1|Aldehyde dehydrogenase family 7 member A1|Alpha AASA dehydrogenase|Alpha aminoadipic semialdehyde dehydrogenase|Alpha-AASA dehydrogenase|Alpha-aminoadipic semialdehyde dehydrogenase|Antiquitin 1|Antiquitin|Antiquitin-1|ATQ1|Betaine aldehyde dehydrogenase|Delta1 piperideine 6 carboxylate dehydrogenease|Delta1-piperideine-6-carboxylate dehydrogenase|EPD|P6c dehydrogenase|PDE
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alpha-aminoadipic semialdehyde dehydrogenase protein (386-515AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ALDH7A1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human