Anti-CDS2 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77852-917
New Product
- Antigen Symbol:CDS2
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O95674
- Antigen Synonyms:CDS2Phosphatidate cytidylyltransferase 2|EC 2.7.7.41|CDP-DAG synthase 2|CDP-DG synthase 2|CDP-diacylglycerol synthase 2|CDS 2|CDP-diglyceride pyrophosphorylase 2|CDP-diglyceride synthase 2|CTP:phosphatidate cytidylyltransferase 2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Phosphatidate cytidylyltransferase 2 protein (354-445AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: CDS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human