Anti-BCL2L11 Rabbit Recombinant Polyclonal Antibody (Biotin)
Catalog # 77851-842
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BCL2L11
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:O43521
- Antigen Synonyms:BCL2 like 11 |B2L11_HUMAN|BAM|Bcl 2 interacting protein Bim|Bcl 2 related ovarian death agonist|Bcl-2-like protein 11|BCL2 interacting mediator of cell death |BCL2 like 11 (apoptosis facilitator)|BCL2 like protein 11 |Bcl2-interacting mediator of cell death|Bcl2-L-11|Bcl2l11|BIM alpha6|BIM|BIM beta6|BIM beta7|BimEL|BimL|BOD
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Bcl-2-like protein 11 protein (1-198AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: BCL2L11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human