Anti-INO80 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: INO80
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-146
- 77845-145
- Antigen Symbol:
- INO80
- INO80
- Antigen Synonyms:
- DNA helicase INO80|hINO80|homolog of yeast INO80|INO80|INO80 complex subunit A|INO80 homolog|INO80_HUMAN|INO80A|INOC1|KIAA1259|Putative DNA helicase INO80 complex homolog 1
- DNA helicase INO80|hINO80|homolog of yeast INO80|INO80|INO80 complex subunit A|INO80 homolog|INO80_HUMAN|INO80A|INOC1|KIAA1259|Putative DNA helicase INO80 complex homolog 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULG1
- Q9ULG1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Chromatin-remodeling ATPase INO80 protein (1365-1495AA)
- Recombinant Human Chromatin-remodeling ATPase INO80 protein (1365-1495AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice