Anti-ACTN3 Rabbit Recombinant Polyclonal Antibody
Catalog # 77850-685
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACTN3
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q08043
- Antigen Synonyms:Actinin alpha 3|actinin|alpha|ACTN 3|ACTN3|ACTN3_HUMAN|Alpha actinin skeletal muscle|Alpha actinin skeletal muscle isoform 3|Alpha-Actinin 3|Alpha-actinin skeletal muscle isoform 3|Alpha-actinin-3|F actin cross linking protein|F-actin cross-linking protein|MGC117002|MGC117005|OTTHUMP00000219049|OTTHUMP00000219054
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Alpha-actinin-3 protein (497-667AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: ACTN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human