Anti-INA Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: INA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77849-473
- 77849-472
- Antigen Symbol:
- INA
- INA
- Antigen Synonyms:
- 66 kDa neurofilament protein|AINX_HUMAN|Alpha Inx|Alpha-internexin|Alpha-Inx|Internexin neuronal intermediate filament protein alpha|MGC12702|NEF 5|NEF5|Neurofilament 5 (66kD)|Neurofilament 5|Neurofilament 66|Neurofilament 66 tax binding protein|Neurofilament-66|NF 66|NF-66|NF66|TXBP 1|TXBP1
- 66 kDa neurofilament protein|AINX_HUMAN|Alpha Inx|Alpha-internexin|Alpha-Inx|Internexin neuronal intermediate filament protein alpha|MGC12702|NEF 5|NEF5|Neurofilament 5 (66kD)|Neurofilament 5|Neurofilament 66|Neurofilament 66 tax binding protein|Neurofilament-66|NF 66|NF-66|NF66|TXBP 1|TXBP1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q16352
- Q16352
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human INA
- Synthetic peptide of Human INA
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice