Anti-ADO Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:10000, WB:1:1000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: ADO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-959
- 77847-958
- Antigen Symbol:
- ADO
- ADO
- Antigen Synonyms:
- 2 aminoethanethiol dioxygenase|2-aminoethanethiol dioxygenase|ADO 2 aminoethanethiol (cysteamine) dioxygenase|ADO|AEDO_HUMAN|C10orf22|Chromosome 10 open reasing frame 22|cysteamine (2-aminoethanethiol) dioxygenase (ADO)|Cysteamine dioxygenase|DKFZp564C046|FLJ14547
- 2 aminoethanethiol dioxygenase|2-aminoethanethiol dioxygenase|ADO 2 aminoethanethiol (cysteamine) dioxygenase|ADO|AEDO_HUMAN|C10orf22|Chromosome 10 open reasing frame 22|cysteamine (2-aminoethanethiol) dioxygenase (ADO)|Cysteamine dioxygenase|DKFZp564C046|FLJ14547
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q96SZ5
- Q96SZ5
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ADO
- Synthetic peptide of Human ADO
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice