Anti-ETFDH Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: ETFDH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
- Catalog No:
- 77736-221
- 77854-954
- Antigen Symbol:
- ETFDH
- ETFDH
- Antigen Synonyms:
- Electron transfer flavoprotein-ubiquinone oxidoreductase|mitochondrial|ETF-QO|ETF-ubiquinone oxidoreductase|EC 1.5.5.1|Electron-transferring-flavoprotein dehydrogenase|ETF dehydrogenase
- Electron transfer flavoprotein-ubiquinone oxidoreductase|mitochondrial|ETF-QO|ETF-ubiquinone oxidoreductase|EC 1.5.5.1|Electron-transferring-flavoprotein dehydrogenase|ETF dehydrogenase
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Pig
- Pig
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55931
- P55931
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Pig Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial protein (34-617AA)
- Recombinant Pig Electron transfer flavoprotein-ubiquinone oxidoreductase, mitochondrial protein (34-617AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice