Anti-MC3R Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
0
Type:
Antigen: MC3R
Clonality: Recombinant polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77742-863
- 77859-557
- Antigen Symbol:
- MC3R
- MC3R
- Antigen Synonyms:
- BMIQ9| MC3| MC3 R| MC3 Receptor| MC3-R| MC3R_HUMAN| Melanocortin 3 receptor| Melanocortin receptor 3| OB20| Obesity quantitative trait locus| OQTL
- BMIQ9|MC3|MC3 R|MC3 Receptor|MC3-R|MC3R_HUMAN|Melanocortin 3 receptor|Melanocortin receptor 3|OB20|Obesity quantitative trait locus|OQTL
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P41968
- P41968
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Melanocortin receptor 3 protein (38-74AA)
- Recombinant Human Melanocortin receptor 3 protein (38-74AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice