Anti-LIM2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: LIM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77858-993
- 77742-186
- Antigen Symbol:
- LIM2
- LIM2
- Antigen Synonyms:
- Lens fiber membrane intrinsic protein|Lens intrinsic membrane protein 2 19kDa|Lens intrinsic membrane protein 2|LIM 2|LMIP_HUMAN|MP17|MP18|MP19|MP20|To3
- Lens fiber membrane intrinsic protein|Lens intrinsic membrane protein 2 19kDa|Lens intrinsic membrane protein 2|LIM 2|LMIP_HUMAN|MP17|MP18|MP19|MP20|To3
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P55344
- P55344
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Lens fiber membrane intrinsic protein (25-110AA)
- Recombinant Human Lens fiber membrane intrinsic protein (25-110AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice