Anti-HLX Rabbit Polyclonal Antibody
Catalog # 77740-127
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:HLX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q14774
- Antigen Synonyms:H2.0 like homeo box 1| H2.0 like homeobox 1| H2.0 like homeobox 1 Drosophila| H2.0-like homeobox protein| HB24| HLX_HUMAN| Homeobox protein HB24| Homeobox protein HLX1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Homeobox protein HLX1
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: HLX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat