Anti-MXD3 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MXD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77860-154
- 77743-677
- Antigen Symbol:
- MXD3
- MXD3
- Antigen Synonyms:
- bHLHc13|Class C basic helix-loop-helix protein 13|FLJ35523|Likely ortholog of mouse Max dimerization protein 3|MAD3_HUMAN|MAX dimerization protein 3|Max dimerizer 3|Max-associated protein 3|Max-interacting transcriptional repressor MAD3|MGC2383|Myx|OTTHUMP00000223596|OTTHUMP00000223598
- bHLHc13|Class C basic helix-loop-helix protein 13|FLJ35523|Likely ortholog of mouse Max dimerization protein 3|MAD3_HUMAN|MAX dimerization protein 3|Max dimerizer 3|Max-associated protein 3|Max-interacting transcriptional repressor MAD3|MGC2383|Myx|OTTHUMP00000223596|OTTHUMP00000223598
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9BW11
- Q9BW11
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Max dimerization protein 3 protein (1-206AA)
- Recombinant Human Max dimerization protein 3 protein (1-206AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice