Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-FAM13A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-FAM13A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-FAM13A Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77736-550
  • 77855-140
  • Antigen Symbol:
  • FAM13A
  • Antigen Synonyms:
  • FAM13A|FAM13A1|KIAA0914Protein FAM13A
  • FAM13A|FAM13A1|KIAA0914Protein FAM13A
  • Clonality:
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Host:
  • Rabbit
  • Gene ID:
  • O94988
  • O94988
  • Isotype:
  • IgG
  • ELISA:
  • Yes
  • Immunogen:
  • Recombinant Human Protein FAM13A protein (1-201AA)
  • Recombinant Human Protein FAM13A protein (1-201AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications