Anti-KCNJ6 Rabbit Polyclonal Antibody
Catalog # 77741-512
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:KCNJ6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Form:Liquid
- Gene ID:P48051
- Antigen Synonyms:inwardly rectifying subfamily J member 6|BIR1|G protein activated inward rectifier potassium channel 2|G protein-activated inward rectifier potassium channel 2|GIRK-2|GIRK2|hiGIRK2|Inward rectifier K(+) channel Kir3.2|Inward rectifier potassium channel KIR3.2|KATP-2|KATP2|KCNJ6|KCNJ6_HUMAN|Kcnj7|KIR3.2|KPLBS|Potassium channel|Potassium channel inwardly rectifying subfamily J member 6|Potassium inwardly rectifying channel subfamily J member 6|Potassium voltage gated channel subfamily J member 6
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Potassium voltage gated channel subfamily J member 6
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: KCNJ6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat