Anti-FCN1 Rabbit Polyclonal Antibody
Catalog # 77737-004
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FCN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:O00602
- Antigen Synonyms:Ficolin-1|Ficolin A|M ficolin|Ficolin alpha|Ficolin-alpha|Collagen/fibrinogen domain containing protein 1|FCN1_HUMAN|Ficolin-A|Ficolin 1|ficolin (collagen/fibrinogen domain containing) 1|M-ficolin|FCNM|Collagen/fibrinogen domain-containing protein 1
- Storage Buffer:PBS, 0.1% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:M-ficolin
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: FCN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human