Anti-FMO4 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
New Product
Type:
Antigen: FMO4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77855-496
- 77737-331
- Antigen Symbol:
- FMO4
- FMO4
- Antigen Synonyms:
- Dimethylaniline monooxygenase [N oxide forming] 4|Dimethylaniline monooxygenase [N-oxide-forming] 4|Dimethylaniline monooxygenase 4|Dimethylaniline oxidase 4|FMO 2|FMO 4|FMO2|FMO4_HUMAN|Hepatic flavin containing monooxygenase 4|Hepatic flavin-containing monooxygenase 4|OTTHUMP00000060454
- Dimethylaniline monooxygenase [N oxide forming] 4|Dimethylaniline monooxygenase [N-oxide-forming] 4|Dimethylaniline monooxygenase 4|Dimethylaniline oxidase 4|FMO 2|FMO 4|FMO2|FMO4_HUMAN|Hepatic flavin containing monooxygenase 4|Hepatic flavin-containing monooxygenase 4|OTTHUMP00000060454
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P31512
- P31512
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human Dimethylaniline monooxygenase [N-oxide-forming] 4 protein (1-300AA)
- Recombinant human Dimethylaniline monooxygenase [N-oxide-forming] 4 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice