Anti-GTF2A2 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: GTF2A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77856-581
- 77739-210
- Antigen Symbol:
- GTF2A2
- GTF2A2
- Antigen Synonyms:
- General transcription factor IIA|General transcription factor IIA subunit 2|2 (12kD subunit)|2|12kDa|HsT18745|T2AG_HUMAN|TF2A2|TFIIA 12|TFIIA|TFIIA gamma|TFIIA p12 subunit|TFIIA-12|TFIIA-gamma|TFIIA12|TFIIAS|Transcription initiation factor IIA gamma chain|Transcription initiation factor IIA subunit 2
- General transcription factor IIA|General transcription factor IIA subunit 2|2 (12kD subunit)|2|12kDa|HsT18745|T2AG_HUMAN|TF2A2|TFIIA 12|TFIIA|TFIIA gamma|TFIIA p12 subunit|TFIIA-12|TFIIA-gamma|TFIIA12|TFIIAS|Transcription initiation factor IIA gamma chain|Transcription initiation factor IIA subunit 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P52657
- P52657
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Transcription initiation factor IIA subunit 2 protein (1-109AA)
- Recombinant human Transcription initiation factor IIA subunit 2 protein (1-109AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice