Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-AKAP14 Rabbit Polyclonal Antibody
Anti-AKAP14 Rabbit Polyclonal Antibody
Catalog # 77735-786
Supplier:  AFG Bioscience
New Product
Anti-AKAP14 Rabbit Polyclonal Antibody
Catalog # 77735-786
Supplier:  AFG Bioscience
Supplier Number:  A20474-50UG
New Product

Specifications

  • Antigen Symbol:
    AKAP14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    Q86UN6
  • Antigen Synonyms:
    PRKA14|AKAP 28|Protein kinase A-anchoring protein 14|A-kinase anchor protein 28 kDa|AKAP28A-kinase anchor protein 14|AKAP-14
  • Storage Buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    PRKA14
  • Purification:
    Antigen affinity purified
  • Packaging:
    Vial

Specifications

About this item

Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:100 to 1:300, IF: 1:200 to 1:1000, ELISA: 1:5000

Type:
Antigen: AKAP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human