Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-IP6K1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-IP6K1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-IP6K1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77741-149
  • 77858-160
  • Antigen Symbol:
  • IP6K1
  • IP6K1
  • Antigen Synonyms:
  • ATP:1D myo inositol hexakisphosphate phosphotransferase|IHPK1|Inositol hexakisphosphate kinase 1|Inositol hexaphosphate kinase 1|InsP6|InsP6 kinase 1|Ip6k1|IP6K1_HUMAN|Itpk6|Pi uptake stimulator|PiUS
  • ATP:1D myo inositol hexakisphosphate phosphotransferase| IHPK1| Inositol hexakisphosphate kinase 1| Inositol hexaphosphate kinase 1| InsP6| InsP6 kinase 1| Ip6k1| IP6K1_HUMAN| Itpk6| Pi uptake stimulator| PiUS
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q92551
  • Q92551
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Inositol hexakisphosphate kinase 1 protein (1-441AA)
  • Recombinant Human Inositol hexakisphosphate kinase 1 protein (1-441AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications