Anti-MMGT1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: MMGT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77743-238
- 77859-838
- Antigen Symbol:
- MMGT1
- MMGT1
- Antigen Synonyms:
- 9630048L06Rik|BC032271|EMC5|Membrane magnesium transporter 1|MMGT1_HUMAN|RGD1566339|RP11-274K13.3|TMEM32|Transmembrane protein 32
- 9630048L06Rik|BC032271|EMC5|Membrane magnesium transporter 1|MMGT1_HUMAN|RGD1566339|RP11-274K13.3|TMEM32|Transmembrane protein 32
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N4V1
- Q8N4V1
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Membrane magnesium transporter 1 protein (66-131AA)
- Recombinant Human Membrane magnesium transporter 1 protein (66-131AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice